Learn More
Medchemexpress LLC Epoetin beta (rhEPO) | 122312-54-3 | ≥95.0% | 60 kDa | 500 U
SDP
Catalog No. 5000436852
Encompass_Preferred
Epoetin beta (rhEPO) is a recombinant form of erythropoietin. It is responsible for the maintenance of erythropoiesis and can be used for anaemia research.
  • Prevents anaemia and enhances the radiosensitivity of solid growing DS-sarcomas in animal models.
  • Used in clinical trials for conditions such as chemotherapy-induced anemia, stillbirth, neonatal respiratory depression, anemia in traumatic brain injury, and anaemia.
Catalog No. 50-004-36852 Supplier Medchemexpress LLC Supplier No. HY114134500U
Epoetin beta (rhEPO) is a recombinant form of erythropoietin. It is responsible for the maintenance of erythropoiesis and can be used for anaemia research.
  • Prevents anaemia and enhances the radiosensitivity of solid growing DS-sarcomas in animal models.
  • Used in clinical trials for conditions such as chemotherapy-induced anemia, stillbirth, neonatal respiratory depression, anemia in traumatic brain injury, and anaemia.