Learn More
MILLER STEPHENSON CHEMICAL COMPANY INC EPON™ 828 Resix, 1 Quart
SDP
Supplier:  MILLER STEPHENSON CHEMICAL COMPANY INC EPON828RESIN.1QT
Encompass_Preferred
EPON™ Resin 828 is an undiluted clear difunctional bisphenol A/epichlorohydrin derived liquid epoxy resin. When cross-linked or hardened with appropriate curing agents, very good mechanical, adhesive, dielectric and chemical resistance properties are obtained. Because of this versatility, EPON Resin 828 has become a standard epoxy resin used in formulation, fabrication and fusion technology.
Catalog No. NC1621311