Learn More
Medchemexpress LLC Epoprostenol sodium | 61849-14-7 | 98.03% | C20H31NaO5 | 1 MG
SDP
Catalog No. 5000432473
Encompass_Preferred
Epoprostenol sodium (Prostaglandin I2) sodium salt is the synthetic form of the natural prostaglandin derivative prostacyclin (prostaglandin I2). It is registered worldwide for the treatment of Pulmonary arterial hypertension (PAH) and is used in pulmonary hypertension and transplantation as a potent inhibitor of platelet aggregation.
  • Registered worldwide for the treatment of pulmonary arterial hypertension (PAH).
  • Used in pulmonary hypertension and transplantation.
  • Potent inhibitor of platelet aggregation.
  • Induces apoptosis in PAH-PASMCs by upregulating the expression of IP receptor and Fas ligand (FasL).
  • Storage at -20°C, sealed, away from moisture.
  • Compound is unstable in solutions; freshly prepared solutions are recommended.
Catalog No. 50-004-32473 Supplier Medchemexpress LLC Supplier No. HYA0126A1MG
Epoprostenol sodium (Prostaglandin I2) sodium salt is the synthetic form of the natural prostaglandin derivative prostacyclin (prostaglandin I2). It is registered worldwide for the treatment of Pulmonary arterial hypertension (PAH) and is used in pulmonary hypertension and transplantation as a potent inhibitor of platelet aggregation.
  • Registered worldwide for the treatment of pulmonary arterial hypertension (PAH).
  • Used in pulmonary hypertension and transplantation.
  • Potent inhibitor of platelet aggregation.
  • Induces apoptosis in PAH-PASMCs by upregulating the expression of IP receptor and Fas ligand (FasL).
  • Storage at -20°C, sealed, away from moisture.
  • Compound is unstable in solutions; freshly prepared solutions are recommended.