Learn More
Aviva Systems Biology Corp Epor Antibody - N-terminal region (ARP44217_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44217_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Epor. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-285-129