Learn More
Aviva Systems Biology Corp EPOR Antibody - N-terminal region (ARP44216_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44216_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EPOR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EPOR. Alternate names: MGC138358, EPO-RApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-58748