Learn More
Cayman Chemical EpothIlon B 10mg
SDP
Catalog No. 502952279
Encompass_Preferred
A macrolide that causes the formation of bundles of intracellular microtubules in non-mitotic cells, induces the formation of hyperstable tubulin polymers, and arrests cell cycling in mitosis; induces mitotic arrest at nanomolar IC50 values in cell lines from ovarian, breast, lung, colon, prostate, and squamous cancer
Catalog No. 50-295-2279 Supplier Cayman Chemical Supplier No. 1092410 MG
A macrolide that causes the formation of bundles of intracellular microtubules in non-mitotic cells, induces the formation of hyperstable tubulin polymers, and arrests cell cycling in mitosis; induces mitotic arrest at nanomolar IC50 values in cell lines from ovarian, breast, lung, colon, prostate, and squamous cancer