Learn More
Selleck Chemical LLC Epothilone A S1297-2mg
SDP
Catalog No. 503108288
Encompass_Preferred
Epothilone A is a paclitaxel-like microtubule-stabilizing agent with EC0 01 of 2 M
Catalog No. 50-310-8288 Supplier Selleck Chemical LLC Supplier No. S12972MG
Epothilone A is a paclitaxel-like microtubule-stabilizing agent with EC0 01 of 2 M