Learn More
Medchemexpress LLC Epothilone B | 152044-54-7 | 99.9% | 507.68 | 1 ML
SDP
Catalog No. 5000384668
Encompass_Preferred
Epothilone B is a microtubule stabilizer (MSA) with a Ki of 0.71μM and possesses blood-brain barrier permeability. It functions by binding to the αβ-tubulin heterodimer subunit, which leads to a decrease in αβ-tubulin dissociation. It is intended for research use only and is not sold to patients.
  • Microtubule stabilizer (MSA)
  • Blood-brain barrier permeability
  • Binds to the αβ-tubulin heterodimer subunit
  • Decreases αβ-tubulin dissociation
Catalog No. 50-003-84668 Supplier Medchemexpress LLC Supplier No. HY1702910MM/1ML
Epothilone B is a microtubule stabilizer (MSA) with a Ki of 0.71μM and possesses blood-brain barrier permeability. It functions by binding to the αβ-tubulin heterodimer subunit, which leads to a decrease in αβ-tubulin dissociation. It is intended for research use only and is not sold to patients.
  • Microtubule stabilizer (MSA)
  • Blood-brain barrier permeability
  • Binds to the αβ-tubulin heterodimer subunit
  • Decreases αβ-tubulin dissociation