Learn More
Apexbio Technology LLC Epothilone B (EPO906, Patupilone)(Synonyms: Patupilone, EPO906, Epothilone B, (-)-Epothilone B, Epothilone B lactam), 25mg, CAS: 152044-54-7.
SDP
Catalog No. 5000060267
Encompass_Preferred
Epothilone B (EPO906 Patupilone CAS 152044-54-7) is a naturally occurring macrocyclic lactone initially isolated from the myxobacterium Sorangium cellulosum Sharing a mechanism of action similar to paclitaxel Epothilone B stabilizes microtubules thereby inhibiting cellular mitosis In vitro Epothilone B displays potent cytotoxic activity with an EC0 01 of approximately 1 8 nM Importantly Epothilone B effectively counters resistance mediated via P-glycoprotein which confers paclitaxel resistance and exhibits superior aqueous solubility compared to paclitaxel Research indicates Epothilone B treatment elevates intracellular free calcium and cytosolic cytochrome C in ovarian cancer SKOV-3 cells highlighting its utility as a research tool in cancer biology and chemotherapy resistance studies
Catalog No. 50-000-60267 Supplier Apexbio Technology LLC Supplier No. A163025
Epothilone B (EPO906 Patupilone CAS 152044-54-7) is a naturally occurring macrocyclic lactone initially isolated from the myxobacterium Sorangium cellulosum Sharing a mechanism of action similar to paclitaxel Epothilone B stabilizes microtubules thereby inhibiting cellular mitosis In vitro Epothilone B displays potent cytotoxic activity with an EC0 01 of approximately 1 8 nM Importantly Epothilone B effectively counters resistance mediated via P-glycoprotein which confers paclitaxel resistance and exhibits superior aqueous solubility compared to paclitaxel Research indicates Epothilone B treatment elevates intracellular free calcium and cytosolic cytochrome C in ovarian cancer SKOV-3 cells highlighting its utility as a research tool in cancer biology and chemotherapy resistance studies