Learn More
Selleck Chemical LLC Patupilone (Epothilone B) 5mg 152044-54-7 EPO906
SDP
Catalog No. 507133
Encompass_Preferred
Patupilone (EPO906, Epothilone B) is a paclitaxel-like microtubule-stabilizing agent with EC0.01 of 1.8 ?M. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-713-3 Supplier Selleck Chemical LLC Supplier No. S13645MG
Patupilone (EPO906, Epothilone B) is a paclitaxel-like microtubule-stabilizing agent with EC0.01 of 1.8 ?M. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.