Learn More
Abcam Epothilone B, Microtubule stabilizing agent, 10UG
Catalog No. 502706732
Encompass_Preferred
MW 507.7 Da, Purity >98%. Microtubule-stabilizing agent. Analog of Epothilone A and D (ab143615 and ab143616). Displays highest efficacy in class III β-tubulin overexpressing malignancies. Inhibits Paclitaxel (ab120143)-resistant cell lines. Shows antitumor effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6732 Supplier Abcam Supplier No. AB14127110UG
MW 507.7 Da, Purity >98%. Microtubule-stabilizing agent. Analog of Epothilone A and D (ab143615 and ab143616). Displays highest efficacy in class III β-tubulin overexpressing malignancies. Inhibits Paclitaxel (ab120143)-resistant cell lines. Shows antitumor effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement