Learn More
Abcam Epothilone D, Microtubule-stabilizing agent, 1MG
Catalog No. 502706104
Encompass_Preferred
MW 491.7 Da. Microtubule-stabilizing agent. Epothilone A (ab143615) and B (ab141271) analog. Improves microtubule density and axonal integrity Shows antineoplastic and neuroprotective effects in vivo. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6104 Supplier Abcam Supplier No. AB1436161MG
MW 491.7 Da. Microtubule-stabilizing agent. Epothilone A (ab143615) and B (ab141271) analog. Improves microtubule density and axonal integrity Shows antineoplastic and neuroprotective effects in vivo. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement