Learn More
Medchemexpress LLC Epothilone F | 208518-52-9 | >99.0% | 25 MG
SDP
Catalog No. 5000341952
Encompass_Preferred
Epothilone F is a 16-membered macrolide microtubule-targeting agent that exhibits significant anticancer activity. It is particularly effective against paclitaxel-resistant cancer cells and has been widely used in cancer-related research. It inhibits the proliferation of breast cancer cells, non-small cell lung cancer cells, and drug-resistant ovarian cancer cells.
  • Microtubule-targeting agent
  • Exhibits significant anticancer activity
  • Effective against paclitaxel-resistant cancer cells
  • Inhibits proliferation of breast cancer cells, non-small cell lung cancer cells, and drug-resistant ovarian cancer cells
Catalog No. 50-003-41952 Supplier Medchemexpress LLC Supplier No. 25MGHY127079
Epothilone F is a 16-membered macrolide microtubule-targeting agent that exhibits significant anticancer activity. It is particularly effective against paclitaxel-resistant cancer cells and has been widely used in cancer-related research. It inhibits the proliferation of breast cancer cells, non-small cell lung cancer cells, and drug-resistant ovarian cancer cells.
  • Microtubule-targeting agent
  • Exhibits significant anticancer activity
  • Effective against paclitaxel-resistant cancer cells
  • Inhibits proliferation of breast cancer cells, non-small cell lung cancer cells, and drug-resistant ovarian cancer cells