Learn More
Medchemexpress LLC Epoxiconazole | 133855-98-8 | 99.0% | 25 MG
SDP
Catalog No. 5000233329
Encompass_Preferred
Epoxiconazole is a triazole-class fungicide and demethylation inhibitor of ergosterol biosynthesis, supplied as a high-purity analytical reagent for laboratory research.
  • Triazole fungicide and ergosterol biosynthesis inhibitor.
  • High purity, 99.0% (as supplied).
  • Chemical formula C17H13ClFN3O; molecular weight 329.76 g/mol.
  • Suitable as a reference standard or reagent for biochemical assays.
  • Available as a small-scale solid dose for analytical testing.
  • Includes datasheet and safety data sheet for handling and storage guidance.
Catalog No. 50-002-33329 Supplier Medchemexpress LLC Supplier No. HY11968325MG
Epoxiconazole is a triazole-class fungicide and demethylation inhibitor of ergosterol biosynthesis, supplied as a high-purity analytical reagent for laboratory research.
  • Triazole fungicide and ergosterol biosynthesis inhibitor.
  • High purity, 99.0% (as supplied).
  • Chemical formula C17H13ClFN3O; molecular weight 329.76 g/mol.
  • Suitable as a reference standard or reagent for biochemical assays.
  • Available as a small-scale solid dose for analytical testing.
  • Includes datasheet and safety data sheet for handling and storage guidance.