Learn More
Medchemexpress LLC Epoxiconazole | 133855-98-8 | 99.0% | 50 MG
SDP
Catalog No. 5000233736
Encompass_Preferred
Epoxiconazole (Standard) is an analytical reference standard of the fungicide epoxiconazole, intended for research and analytical applications such as qualitative and quantitative methods by HPLC, GC, and MS.
  • Analytical standard for HPLC, GC, and MS methods.
  • Used as a reference for qualitative and quantitative analyses.
  • Molecular formula C17H13ClFN3O and molecular weight 329.76 g/mol.
  • Available in small pack sizes suitable for laboratory reference use.
Catalog No. 50-002-33736 Supplier Medchemexpress LLC Supplier No. HY11968350MG
Epoxiconazole (Standard) is an analytical reference standard of the fungicide epoxiconazole, intended for research and analytical applications such as qualitative and quantitative methods by HPLC, GC, and MS.
  • Analytical standard for HPLC, GC, and MS methods.
  • Used as a reference for qualitative and quantitative analyses.
  • Molecular formula C17H13ClFN3O and molecular weight 329.76 g/mol.
  • Available in small pack sizes suitable for laboratory reference use.