Learn More
Medchemexpress LLC Epoxidized soya bean 10g
SDP
Catalog No. 5000205926
Encompass_Preferred
Epoxidized soya bean oil (ESBO) is a vegetable oil-derived organic compound used as a plasticizer and stabilizer in various applications It is produced by epoxidation of soybean oil which introduces epoxy groups into the fatty acid chains of the oil ESBO is a viscous pale yellow liquid that is soluble in many organic solvents such as chloroform and ethanol but insoluble in water It is commonly used as a plasticizer in polyvinyl chloride (PVC) products including toys food packaging materials and medical devices In addition to its plasticizing properties ESBO acts as an antioxidant and UV stabilizer helping to prevent degradation and discoloration of PVC products over time ESBOs have been investigated for their potential use in biodegradable plastics and as bio-based alternatives to traditional petroleum-derived plasticizers
Catalog No. 50-002-05926 Supplier Medchemexpress LLC Supplier No. HYW09328210G
Add to Cart
Add to Cart
Epoxidized soya bean oil (ESBO) is a vegetable oil-derived organic compound used as a plasticizer and stabilizer in various applications It is produced by epoxidation of soybean oil which introduces epoxy groups into the fatty acid chains of the oil ESBO is a viscous pale yellow liquid that is soluble in many organic solvents such as chloroform and ethanol but insoluble in water It is commonly used as a plasticizer in polyvinyl chloride (PVC) products including toys food packaging materials and medical devices In addition to its plasticizing properties ESBO acts as an antioxidant and UV stabilizer helping to prevent degradation and discoloration of PVC products over time ESBOs have been investigated for their potential use in biodegradable plastics and as bio-based alternatives to traditional petroleum-derived plasticizers