Learn More
Apexbio Technology LLC Epoxomicin 134381-21-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062942
Encompass_Preferred
Epoxomicin (CAS 134381-21-8) is a naturally occurring proteasome inhibitor initially isolated from actinomycete cultures It acts primarily by forming covalent bonds through its -epoxyketone moiety with catalytic residues of the proteasome resulting in potent inhibition of the chymotrypsin-like (CTRL) activity of the 20S proteasome subunit Epoxomicin also inhibits proteasomal trypsin-like and peptidyl-glutamyl peptide hydrolysis activities albeit at significantly lower rates It exhibits anti-inflammatory and antitumor activities and is employed experimentally to study ubiquitin-proteasome-mediated cellular pathways bone formation regulation and Parkinson s disease model generation
Catalog No. 50-000-62942 Supplier Apexbio Technology LLC Supplier No. A26065.1
Epoxomicin (CAS 134381-21-8) is a naturally occurring proteasome inhibitor initially isolated from actinomycete cultures It acts primarily by forming covalent bonds through its -epoxyketone moiety with catalytic residues of the proteasome resulting in potent inhibition of the chymotrypsin-like (CTRL) activity of the 20S proteasome subunit Epoxomicin also inhibits proteasomal trypsin-like and peptidyl-glutamyl peptide hydrolysis activities albeit at significantly lower rates It exhibits anti-inflammatory and antitumor activities and is employed experimentally to study ubiquitin-proteasome-mediated cellular pathways bone formation regulation and Parkinson s disease model generation