Learn More
AdipoGen Epoxomicin (250 µg)
SDP
Supplier:  AdipoGen AGCN20422C250
Encompass_Preferred
Epoxomicin (250 µg), AdipoGen Life Sciences. Chemical. CAS: 134381-21-8. Formula: C28H50N4O7. MW: 554.7. Synthetic. Antibiotic. Potent anticancer compound. Cell permeable, potent, selective and irreversible 20S proteasome inhibitor. Predominantly inhibits the chymotrypsin-like (CTRL) activity of the proteasome. Exhibits lower level inhibition of proteasome trypsin-like and caspase-like activitives (100 and 1,000-fold slower rates respectively). Anti-inflammatory. Antimicrobial and antimalarial. Anti-parasitic. Stimulates bone formation by inhibiting osteoblast proteasome activity. Induces Parkinson's-like symptoms in rats. The ubiquitin-proteasome system (UPS) and autophagy serve as two complementary, reciprocally regulated protein degradation systems. Blockade of UPS by Epoxomicin activates autophagy.
Catalog No. 50-148-9981