Learn More
Apexbio Technology LLC Epoxomicin, 5mg. Cas: 134381-21-8 MFCD: MFCD03791061. Proteasome inhibitor
SDP
Supplier:  Apexbio Technology LLC A26065
Encompass_Preferred
Epoxomicin was originally isolated from the culture medium of an Actinomycetes strain based on its in vivo antitumor activity against murine B16 melanoma. Epoxomicin is a naturally occurring selective proteasome inhibitor with anti-inflammatory activity. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4754