Learn More
Enzo Life Sciences Epoxomicin, (100µg), CAS Number: 134381-21-8
SDP
Catalog No. NC9982514
Encompass_Preferred
Key inhibitor for use in proteasome research. Formula: C28H50N4O7. MW: 554.7. CAS Number: 134381-21-8. MI: 14: 3630. Purity: ≥95% (HPLC). Appearance: Lyophilized. Very thin transparent film. Solubility: Soluble in DMSO (15mg/ml) or dichloromethane:methanol (9:1); insoluble in water. Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C
Catalog No. NC9982514 Supplier Enzo Life Sciences Supplier No. BMLPI1270100
Key inhibitor for use in proteasome research. Formula: C28H50N4O7. MW: 554.7. CAS Number: 134381-21-8. MI: 14: 3630. Purity: ≥95% (HPLC). Appearance: Lyophilized. Very thin transparent film. Solubility: Soluble in DMSO (15mg/ml) or dichloromethane:methanol (9:1); insoluble in water. Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C