Learn More
APPLIED INDUSTRIAL TECHNOLOGY EPOXY, 5-MINUTE, DEVCON
Catalog No. NC9863515
Encompass_Preferred
5 Minute Epoxy,Rapid-curing, general-purpose adhesive/encapsulant,size:25ml Dev-Tube.Devcon,Bonds rigid durable substrates such as metals, glass, ceramics, concrete, and wood in all combinations,Forms a clear hard rigid bond or coating in m
Catalog No. NC9863515 Supplier APPLIED INDUSTRIAL TECHNOLOGY Supplier No. 14250
5 Minute Epoxy,Rapid-curing, general-purpose adhesive/encapsulant,size:25ml Dev-Tube.Devcon,Bonds rigid durable substrates such as metals, glass, ceramics, concrete, and wood in all combinations,Forms a clear hard rigid bond or coating in m