Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Epoxy activated Magnetic Beads
SCI_ED

Catalog No. S001128
Change view
Click to view available options
Capacity (Metric):
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S001128 1 mL
S001158 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S001128 Supplier G-Biosciences Supplier No. 7862214
Add to Cart
Add to Cart

Epoxy Activated Magnetic Beads are suitable to couple biomolecules with free amine (NH2) and thiol (SH) groups.

  • Usage: Coupling biomolecules to active Epoxy Groups
  • Specificity: Exposed amine (NH2) or thiol (SH) groups on biomolecule
  • Bead Ligand: Chemically active Epoxy groups Epoxy
  • Group density: 50 mg/mL
  • Bead size: 300 nm
  • Matrix: Polymer
  • Storage: 4°C

Specifications

Capacity (Metric) 1 mL
Particle Size 150 to 300 μm
Form Resin Slurry
Product Line DynaMag
For Use With (Application) Coupling biomolecules with free amine (NH2) and thiol (SH) groups.
Type Magnetic Affinity Bead
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.