Learn More
G-Biosciences Epoxy activated Magnetic Beads

Description
- Usage: Coupling biomolecules to active Epoxy Groups
- Specificity: Exposed amine (NH2) or thiol (SH) groups on biomolecule
- Bead Ligand: Chemically active Epoxy groups Epoxy
- Group density: 50 mg/mL
- Bead size: 300 nm
- Matrix: Polymer
- Storage: 4°C
Specifications
Specifications
| Particle Size | 150 to 300 μm |
| Form | Resin Slurry |
| Product Line | DynaMag |
| For Use With (Application) | Coupling biomolecules with free amine (NH2) and thiol (SH) groups. |
| Type | Magnetic Affinity Bead |
| Capacity (Metric) | 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.