Learn More
Bioworld EPOXY-ACTIVATED SEPAROPORE
Supplier:  Bioworld 201810642
Encompass
EPOXY-ACTIVATED SEPAROPORE (AGAROSE) 6B-CL: Epoxy-activated Separopore 6B-CL Used in coupling of hydroxy, amino or thiol groups to Separopore 6B via a 12-carbon hydrophilic spacer arm.; Pre-activated medium for immobilization of ligands ...
Catalog No. 50-197-829