Learn More
Ace Glass, Inc. EPOXY ADHESIVE 30ML JAR EPOXY ADHESIVE 30ML JAR
SDP
Supplier:  Ace Glass, Inc. 756010
Encompass_Preferred
A two-component system; cures to a film highly resistant to chemicals which tightly bonds the PTFE sleeve to glass. If sleeve is damaged, the epoxy may be removed with boiling formic acid and a new sleeve may be attached. Supplied in 30mL jars with activator in separate container. 30mL will cement 25 or more joints, depending on size
Catalog No. NC9083033