Learn More
Marketlab, Inc. Epoxy-coated Steel Wire Benchtop Biohazard Bag Holder
Description
- Rectangular steel wire frame fits Benchtop Biohazard Bags or 8.5 in. x 11 in. bags
- Ideal for collecting pipette tips
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Orange |
| Disposable | Reusable |
| Material | Epoxy-coated Wire |
| Width (English) | 5 in. |
| Product Type | Biohazard Bag Holder |
| Width (Metric) | 12.8 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.