Learn More
MSC Bon Tool Plastic Loop Roller, Coarse, 18 in, 1 ea
Catalog No. 502819440
Encompass
This specially designed plastic looped nap roller is intended for applying epoxy and other coatings to concrete surfaces.
  • Specially designed plastic looped nap
  • Phenolic core
  • Length of 18 in
  • End caps sold separately
Catalog No. 50-281-9440 Supplier MSC Supplier No. 29505955
This specially designed plastic looped nap roller is intended for applying epoxy and other coatings to concrete surfaces.
  • Specially designed plastic looped nap
  • Phenolic core
  • Length of 18 in
  • End caps sold separately