Learn More
Electron Microscopy Sciences Cold Mount Epoxy Dye 20 GR
SDP
Catalog No. 50980292
Encompass_Preferred
Fluorescent dye for use in microscopy for cold mounting epoxies including EpoFix and Unimount to make cracks and pores visible.
Catalog No. 50-980-292 Supplier Electron Microscopy Sciences Supplier No. 1245
Fluorescent dye for use in microscopy for cold mounting epoxies including EpoFix and Unimount to make cracks and pores visible.