Learn More
Mitchell Plastics Side Loading Powder-Coated Stainless Steel Glove Box Holder
Catalog No. 06666818
Change view
Click to view available options
Capacity:
1 Box
2 Boxes
3 Boxes
Color:
Blue
White
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Color
06-666-818 3 Boxes Blue
06-666-816 1 Box Blue
06-666-817 2 Boxes Blue
06-666-811 2 Boxes White
06-666-812 3 Boxes White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 06-666-818 Supplier Mitchell Plastics Supplier No. MGPCLL3000B
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Stainless steel holders can be cleaned with bleach and other disfectants, making them ideal for use in clean rooms or other areas that require sterilization.
  • Can be mounted horizontally or vertically
  • Pre-drilled holes and double-sided tape are included for wall mounting
Specifications
Product Type Glove Box Holder
Color Blue
Dimensions (L x W x H) 4 x 31.5 x 6.75 in.
Holds Disposable Glove Boxes
Material Powder-coated Steel
Mounting Keyhole Slots
No. of Compartments 1
Product Style Side Loading
Capacity 3 Boxes
Includes Foam tape