Learn More
Grainger Eclectic Products Amazing Goop Epoxy Sealer, Gray, Solvent, 8 lb
Catalog No. 502763740
Encompass_Preferred
This epoxy sealer is a solvent-based, gray resin coating designed for effective sealing applications. It provides a durable finish with resistance to abrasion, flaking, and water.
  • Provides 80 sq ft of coverage
  • Dry and cure time of 10 to 12 hr
  • Compatible with fiberglass, metal, and wood surfaces
  • Suitable for application via foam roller, brush, squeegee, or toothed trowel
  • UV resistant formula
  • Designed for application in temperatures of 65°F to 70°F
Catalog No. 50-276-3740 Supplier Grainger Supplier No. 16NG94
This epoxy sealer is a solvent-based, gray resin coating designed for effective sealing applications. It provides a durable finish with resistance to abrasion, flaking, and water.
  • Provides 80 sq ft of coverage
  • Dry and cure time of 10 to 12 hr
  • Compatible with fiberglass, metal, and wood surfaces
  • Suitable for application via foam roller, brush, squeegee, or toothed trowel
  • UV resistant formula
  • Designed for application in temperatures of 65°F to 70°F