Learn More
Electron Microscopy Sciences Epoxy Tissue Stain, 100 mL
SDP
Catalog No. 50980450
Encompass_Preferred
Toluidine Blue and Basic Fuchsin. For staining semi thin sections of epoxy embedded tissues.
Catalog No. 50-980-450 Supplier Electron Microscopy Sciences Supplier No. 14950
Toluidine Blue and Basic Fuchsin. For staining semi thin sections of epoxy embedded tissues.