Learn More
Uline Epoxy Wire Shelving Unit, 48 x 18 x 72"", Green
Catalog No. NC1880184
Encompass
Coated shelving provides superior chemical resistance. Recommended for wet environments, labs and kitchen coolers. Epoxy coating withstands harsh cleaning, salt and water. Shelves adjust in 1" increments.
Catalog No. NC1880184 Supplier Uline Supplier No. H4306G
Coated shelving provides superior chemical resistance. Recommended for wet environments, labs and kitchen coolers. Epoxy coating withstands harsh cleaning, salt and water. Shelves adjust in 1" increments.