Learn More
Microsolv Technology Corporation Epp Am Vial/Cap Aq Preslit Si
Catalog No. 5001452298
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Amber Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE, Pre-Slit Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52298 Supplier Microsolv Technology Corporation Supplier No. 9509S3WAPAQ
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Amber Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE, Pre-Slit Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.