Learn More
Microsolv Technology Corporation Epp Clear Vial And Blue Screw
Catalog No. 5001452110
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Silicone Rubber / PTFE Septa, bonded in the Blue Caps, MicroSolv Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52110 Supplier Microsolv Technology Corporation Supplier No. 9502S1CP
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Silicone Rubber / PTFE Septa, bonded in the Blue Caps, MicroSolv Brand Easy Purchase Pack. 1/PK.