Learn More
Microsolv Technology Corporation Epp Clear Vial And Screw Caps
Catalog No. 5001452120
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Red PTFE / White Silicone Rubber / Red PTFE Septa, bonded in the Blue Caps, MicroSolv Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52120 Supplier Microsolv Technology Corporation Supplier No. 9502S2CP
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Red PTFE / White Silicone Rubber / Red PTFE Septa, bonded in the Blue Caps, MicroSolv Brand Easy Purchase Pack. 1/PK.