Learn More
Microsolv Technology Corporation Epp Clear Vials And Snap Cap
Catalog No. 5001451957
Encompass_Preferred
Vial & Cap Kit includes 100 EA 2ml, Snap Top, Clear Autosampler Vials and 100 EA matching Snap Caps with Silicone Rubber / PTFE Pre-Slit Septa fitted in the Blue Caps. MicroSolv Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-51957 Supplier Microsolv Technology Corporation Supplier No. 9502C3CP
Vial & Cap Kit includes 100 EA 2ml, Snap Top, Clear Autosampler Vials and 100 EA matching Snap Caps with Silicone Rubber / PTFE Pre-Slit Septa fitted in the Blue Caps. MicroSolv Brand Easy Purchase Pack. 1/PK.