Learn More
Microsolv Technology Corporation Epp Clear Write On Vial Cap
Catalog No. 5001451959
Encompass_Preferred
Vial & Cap Kit includes 100 EA 2ml, Snap Top, Clear Autosampler Vials with Write-On Patch and 100 EA matching Snap Caps with Silicone Rubber / PTFE Pre-Slit, Septa fitted in the Blue Caps. MicroSolv Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-51959 Supplier Microsolv Technology Corporation Supplier No. 9502C3WCP
Vial & Cap Kit includes 100 EA 2ml, Snap Top, Clear Autosampler Vials with Write-On Patch and 100 EA matching Snap Caps with Silicone Rubber / PTFE Pre-Slit, Septa fitted in the Blue Caps. MicroSolv Brand Easy Purchase Pack. 1/PK.