Learn More
Microsolv Technology Corporation Epp Clear Writeon Vials Cap
Catalog No. 5001451936
Encompass_Preferred
Vial & Cap Kit includes 100 EA 2ml, Snap Top, Clear Autosampler Vials with Write-On Patch and 100 EA matching Snap Caps with Silicone Rubber / PTFE Septa fitted in the Blue Caps. MicroSolv Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-51936 Supplier Microsolv Technology Corporation Supplier No. 9502C1WCP
Vial & Cap Kit includes 100 EA 2ml, Snap Top, Clear Autosampler Vials with Write-On Patch and 100 EA matching Snap Caps with Silicone Rubber / PTFE Septa fitted in the Blue Caps. MicroSolv Brand Easy Purchase Pack. 1/PK.