Learn More
Microsolv Technology Corporation Epp Fi Vial Screw Sil/Ptfe Amb
Catalog No. 5001452378
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 300ul, Fused Insert, Screw Top, Amber Autosampler Vials with Write on Patch and fill lines and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52378 Supplier Microsolv Technology Corporation Supplier No. 9532S1AP
Vial & Cap Kit Includes 100 EA 300ul, Fused Insert, Screw Top, Amber Autosampler Vials with Write on Patch and fill lines and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.