Learn More
Microsolv Technology Corporation Epp Fi Vial Screw Sil/Ptfe Dea
Catalog No. 5001452382
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 300ul, Fused Insert, Screw Top, Clear Deactivated Autosampler Vials with Write on Patch and fill lines and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52382 Supplier Microsolv Technology Corporation Supplier No. 9532S1DCP
Vial & Cap Kit Includes 100 EA 300ul, Fused Insert, Screw Top, Clear Deactivated Autosampler Vials with Write on Patch and fill lines and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.