Learn More
Microsolv Technology Corporation Epp Fi Vial Scrw Sil/Ptslit Am
Catalog No. 5001452383
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 300ul, Fused Insert, Screw Top, Amber Autosampler Vials with Write on Patch and fill lines and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE, Pre-Slit Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52383 Supplier Microsolv Technology Corporation Supplier No. 9532S3AP
Vial & Cap Kit Includes 100 EA 300ul, Fused Insert, Screw Top, Amber Autosampler Vials with Write on Patch and fill lines and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE, Pre-Slit Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.