Learn More
Microsolv Technology Corporation Epp Fi Vialscrwsil/Ptfe Amdeac
Catalog No. 5001452381
Encompass_Preferred
This item has a minimum order quantity of 20 kits per supplier requirements.
Silicone Rubber / White PTFE Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52381 Supplier Microsolv Technology Corporation Supplier No. 9532S1DAP
This item has a minimum order quantity of 20 kits per supplier requirements.
Silicone Rubber / White PTFE Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.