Learn More
Microsolv Technology Corporation Epp Maxrec Vial Clr Prslt Cps
Catalog No. 5001452335
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 1.8ml, Maximum Recovery, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE, Pre-Slit Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52335 Supplier Microsolv Technology Corporation Supplier No. 9512S3CP
Vial & Cap Kit Includes 100 EA 1.8ml, Maximum Recovery, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE, Pre-Slit Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.