Learn More
Microsolv Technology Corporation Epp Mrq Vial/Scrwcap Rsa Pre
Catalog No. 5001452339
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 1.2ml, MRQ, Screw Top, Clear RSA™ Autosampler Vials and 100 EA matching Screw Caps with Clear AQR Silicone Rubber / Clear PTFE, ultra-pure, Pre-Slit Septa, fitted in the Light Blue Caps. RSA Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52339 Supplier Microsolv Technology Corporation Supplier No. 9512S3MPRS
Vial & Cap Kit Includes 100 EA 1.2ml, MRQ, Screw Top, Clear RSA™ Autosampler Vials and 100 EA matching Screw Caps with Clear AQR Silicone Rubber / Clear PTFE, ultra-pure, Pre-Slit Septa, fitted in the Light Blue Caps. RSA Brand Easy Purchase Pack. 1/PK.