Learn More
Microsolv Technology Corporation Epp Vial/Scrwcap Aq Preslit Si
Catalog No. 5001452297
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE, Pre-Slit Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52297 Supplier Microsolv Technology Corporation Supplier No. 9509S3CPAQ
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE, Pre-Slit Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.