Learn More
Microsolv Technology Corporation Epp Vials Wo Amber Lc Clean
Catalog No. 5001452082
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Amber Deactivated Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Clear Silicone Rubber / Clear Tefzel, ultra-pure Septa, fitted in the Blue Caps. MicroSolv Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52082 Supplier Microsolv Technology Corporation Supplier No. 9502S06SWAP
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Amber Deactivated Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Clear Silicone Rubber / Clear Tefzel, ultra-pure Septa, fitted in the Blue Caps. MicroSolv Brand Easy Purchase Pack. 1/PK.