Learn More
Microsolv Technology Corporation Epp Vials Wo Amber Screw Cap
Catalog No. 5001452095
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Amber Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Silicone Rubber / PTFE Septa, fitted in the Robo Style, Blue Caps. MicroSolv Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52095 Supplier Microsolv Technology Corporation Supplier No. 9502S0WAP
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Amber Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Silicone Rubber / PTFE Septa, fitted in the Robo Style, Blue Caps. MicroSolv Brand Easy Purchase Pack. 1/PK.