Learn More
Microsolv Technology Corporation Epp Vials Wo Clear Screw
Catalog No. 5001452084
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Clear Silicone Rubber / Clear Tefzel, ultra-pure Septa, fitted in the Blue Caps. MicroSolv Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52084 Supplier Microsolv Technology Corporation Supplier No. 9502S06WCP
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Clear Silicone Rubber / Clear Tefzel, ultra-pure Septa, fitted in the Blue Caps. MicroSolv Brand Easy Purchase Pack. 1/PK.