Learn More
Microsolv Technology Corporation Epp Wo Amb Vial/Scrwcap Rsa
Catalog No. 5001452292
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Amber RSA™ Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Clear AQR Silicone Rubber / Clear PTFE, ultra-pure Septa, fitted in the Light Blue Caps. RSA Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52292 Supplier Microsolv Technology Corporation Supplier No. 9509S1WAPRS
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Amber RSA™ Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Clear AQR Silicone Rubber / Clear PTFE, ultra-pure Septa, fitted in the Light Blue Caps. RSA Brand Easy Purchase Pack. 1/PK.