Learn More
Microsolv Technology Corporation Epp Wo Amvial/Cap Aq Sil/Ptfe
Catalog No. 5001452291
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Amber Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52291 Supplier Microsolv Technology Corporation Supplier No. 9509S1WAPAQ
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Amber Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE Septa bonded in the Blue Caps. AQ Brand Easy Purchase Pack. 1/PK.